The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159497.1 | NP_001152969.1 | 5 - 6 | 1292 | 91 |
GenBank mRNA | AK076923.1 | - | 5 - 6 | 1300 | 91 |
D45860.1 | - | 5 - 6 | 1164 | 91 |