Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clip4
Entrez Gene ID
78785
Gene Name
CAP-GLY domain containing linker protein family, member 4
Gene Aliases
1700024K14Rik, 1700074B05Rik, 4833417L20Rik, 5830409B12Rik, Rsnl2
Chromosome Location
Chr.17: 71769669 - 71864210 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271484.1 NP_001258413.1 1 - 2 217 83
NM_175378.1 NP_780587.1 1 - 2 217 83
NR_073185.1 1 - 2 217 83
GenBank mRNA AK018895.1 - 1 - 2 210 83
AK029962.1 - 1 - 2 218 83
AK075758.1 - 1 - 2 208 83
AK161301.1 - 1 - 2 207 83

View Full Product Details