Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Galnt14
Entrez Gene ID
71685
Gene Name
UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14
Gene Aliases
0610033M06Rik
Chromosome Location
Chr.17: 73493207 - 73710461 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027864.2 NP_082140.2 5 - 6 926 100
GenBank mRNA AK002747.1 - 5 - 6 898 100
AK049598.1 - 5 - 6 872 100
AK078292.1 - 5 - 6 539 100
BC117800.1 - 5 - 6 710 100

View Full Product Details