The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027166.5 | NP_081442.1 | 2 - 3 | 252 | 70 |
GenBank mRNA | AB098744.1 | - | 1 - 2 | 140 | 70 |
AK010201.1 | - | 2 - 3 | 218 | 70 | |
AK046281.1 | - | 2 - 3 | 251 | 70 | |
AK049096.1 | - | 2 - 3 | 253 | 70 | |
AK049644.1 | - | 2 - 3 | 251 | 70 | |
AK137446.1 | - | 2 - 3 | 224 | 70 | |
AK154461.1 | - | 2 - 3 | 291 | 70 | |
AK165943.1 | - | 2 - 3 | 254 | 70 | |
AK167998.1 | - | 2 - 3 | 268 | 70 | |
AK169241.1 | - | 2 - 3 | 255 | 70 | |
AK169986.1 | - | 2 - 3 | 226 | 70 | |
AK171010.1 | - | 2 - 3 | 253 | 70 | |
AK217637.1 | - | 1 - 2 | 166 | 70 | |
BC085109.1 | - | 2 - 3 | 271 | 70 |