Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thoc1
Entrez Gene ID
225160
Gene Name
THO complex 1
Gene Aliases
3110002N20Rik, AW107452, NMP-84
Chromosome Location
Chr.18: 9958180 - 9995484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153552.3 NP_705780.1 18 - 19 1457 103
GenBank mRNA AK031785.1 - 18 - 19 1458 103
AK032200.1 - 18 - 19 1458 103
AK052835.1 - 18 - 19 1453 103
AK167374.1 - 18 - 19 1471 103
AK168619.1 - 18 - 19 1723 103
BC024951.1 - 18 - 19 1629 103
BC027149.1 - 11 - 12 1236 103

View Full Product Details