The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026484.3 | NP_080760.2 | 1 - 2 | 632 | 85 |
GenBank mRNA | AK014201.1 | - | 1 - 2 | 637 | 85 |
AK017493.1 | - | 1 - 2 | 464 | 85 | |
AK028696.1 | - | 1 - 2 | 633 | 85 | |
AK076556.1 | - | 1 - 2 | 162 | 85 | |
BC023321.1 | - | 2 - 3 | 550 | 85 |