Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cul2
Entrez Gene ID
71745
Gene Name
cullin 2
Gene Aliases
1300003D18Rik, 4932411N15Rik, AI327301, mKIAA4106
Chromosome Location
Chr.18: 3382975 - 3436700 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029402.3 NP_083678.2 17 - 18 1876 100
GenBank mRNA AK004875.2 - 17 - 18 1832 100
AK016520.1 - 17 - 18 1846 100
AK028589.1 - 17 - 18 1845 100
AK144693.1 - 17 - 18 1850 100
AK160597.1 - 17 - 18 1861 100
AK220287.1 - 17 - 18 1851 100
BC025902.1 - 9 - 10 942 100
BC026779.1 - 17 - 18 1777 100
BC027428.1 - 17 - 18 1839 100

View Full Product Details