Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sos1
Entrez Gene ID
20662
Gene Name
son of sevenless homolog 1 (Drosophila)
Gene Aliases
4430401P03Rik, 9630010N06, AI449023
Chromosome Location
Chr.17: 80393752 - 80480453 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009231.2 NP_033257.2 15 - 16 3033 147
GenBank mRNA AK140310.1 - 15 - 16 3033 147

View Full Product Details