The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027236.2 | NP_081512.1 | 2 - 3 | 618 | 118 |
GenBank mRNA | AK008070.1 | - | 2 - 3 | 428 | 118 |
AK031807.1 | - | 2 - 3 | 452 | 118 | |
AK153194.1 | - | 2 - 3 | 449 | 118 | |
AK158977.1 | - | 2 - 3 | 436 | 118 | |
AK168102.1 | - | 2 - 3 | 422 | 118 | |
AK168253.1 | - | 2 - 3 | 422 | 118 | |
AK168598.1 | - | 2 - 3 | 445 | 118 | |
BC024640.1 | - | 2 - 3 | 618 | 118 |