Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Naaladl1
Entrez Gene ID
381204
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 1
Gene Aliases
Gm964, Naaladasel
Chromosome Location
Chr.19: 6105798 - 6115555 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009546.1 NP_001009546.1 7 - 8 1091 68

View Full Product Details