The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164480.1 | NP_001157952.1 | 9 - 10 | 2281 | 69 |
NM_001164481.1 | NP_001157953.1 | 10 - 11 | 2535 | 69 | |
NM_001164482.1 | NP_001157954.1 | 8 - 9 | 2220 | 69 | |
NM_001164568.1 | NP_001158040.1 | 8 - 9 | 2250 | 69 | |
NM_011379.4 | NP_035509.4 | 9 - 10 | 2504 | 69 | |
GenBank mRNA | AK028151.1 | - | 8 - 9 | 1491 | 69 |
AK137578.1 | - | 9 - 10 | 2227 | 69 | |
AK149669.1 | - | 9 - 10 | 2227 | 69 | |
AK156280.1 | - | 10 - 11 | 2489 | 69 | |
AK169945.1 | - | 8 - 9 | 2166 | 69 | |
AK170246.1 | - | 8 - 9 | 2181 | 69 | |
AK220472.1 | - | 9 - 10 | 2417 | 69 | |
BC043679.1 | - | 1 - 2 | 641 | 69 | |
BC054824.1 | - | 9 - 10 | 2225 | 69 | |
D11374.1 | - | 8 - 9 | 2178 | 69 |