Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Syvn1
Entrez Gene ID
74126
Gene Name
synovial apoptosis inhibitor 1, synoviolin
Gene Aliases
1200010C09Rik, AW211966, C85322, D530017H19Rik, Hrd1
Chromosome Location
Chr.19: 6045030 - 6053718 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164709.1 NP_001158181.1 11 - 12 1310 108
NM_028769.5 NP_083045.4 11 - 12 1677 108
GenBank mRNA AK004688.2 - 11 - 12 1164 108
BC042199.1 - 11 - 12 1172 108
BC046829.1 - 11 - 12 1310 108
BC057917.1 - 11 - 12 1591 108
BC080722.1 - 11 - 12 1195 108

View Full Product Details