Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il1rl1
Entrez Gene ID
17082
Gene Name
interleukin 1 receptor-like 1
Gene Aliases
DER4, Fit-1, Ly84, ST2L, St2, St2-rs1, T1, T1/ST2
Chromosome Location
Chr.1: 40404864 - 40465418 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025602.3 NP_001020773.1 7 - 8 1191 148
NM_001294171.1 NP_001281100.1 7 - 8 1024 148
NM_010743.3 NP_034873.2 7 - 8 1191 148
GenBank mRNA AK033417.1 - 7 - 8 1044 148
BC051445.1 - 7 - 8 1001 148
D13695.1 - 6 - 7 883 148
M24843.1 - 7 - 8 1032 148
Y07519.1 - 7 - 8 1035 148

View Full Product Details