Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mdm1
Entrez Gene ID
17245
Gene Name
transformed mouse 3T3 cell double minute 1
Gene Aliases
Arrd2, Mdm-1
Chromosome Location
Chr.10: 118141537 - 118168999 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA M20823.1 - 12 - 13 1978 69

View Full Product Details