The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007935.2 | NP_031961.1 | 13 - 14 | 2462 | 61 |
NM_027497.3 | NP_081773.1 | 13 - 14 | 2613 | 61 | |
GenBank mRNA | AF079765.1 | - | 13 - 14 | 2441 | 61 |
AK040254.1 | - | 13 - 14 | 2613 | 61 | |
AK148194.1 | - | 13 - 14 | 2431 | 61 | |
AK149317.1 | - | 10 - 11 | 1813 | 61 | |
BC138622.1 | - | 13 - 14 | 2441 | 61 | |
BC138623.1 | - | 13 - 14 | 2441 | 61 |