Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdpcp
Entrez Gene ID
216560
Gene Name
WD repeat containing planar cell polarity effector
Gene Aliases
AV249152
Chromosome Location
Chr.11: 21572281 - 21898687 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145425.3 NP_663400.2 9 - 10 846 64
GenBank mRNA AF424701.1 - 9 - 10 809 64
AK043672.1 - 8 - 9 733 64
AK083042.1 - 9 - 10 847 64
BC141166.1 - 9 - 10 810 64

View Full Product Details