Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd39
Entrez Gene ID
109346
Gene Name
ankyrin repeat domain 39
Gene Aliases
9130416N05Rik, C030004B10Rik
Chromosome Location
Chr.1: 36538173 - 36547440 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026241.3 NP_080517.1 3 - 4 424 73
GenBank mRNA AK018681.1 - 3 - 4 425 73
AK030874.1 - - 620 73
AK042896.1 - 3 - 4 425 73
AK081207.1 - 3 - 4 617 73
AK158215.1 - 3 - 4 619 73
AK160231.1 - 3 - 4 440 73
BC117004.1 - 3 - 4 454 73
BC119100.1 - 3 - 4 454 73
BC144893.1 - 3 - 4 108

View Full Product Details