Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd39
Entrez Gene ID
109346
Gene Name
ankyrin repeat domain 39
Gene Aliases
9130416N05Rik, C030004B10Rik
Chromosome Location
Chr.1: 36538173 - 36547440 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK030874.1 - - 312 74
AK081207.1 - 1 - 2 309 74
AK158215.1 - 1 - 2 311 74

View Full Product Details