Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ahsa2
Entrez Gene ID
268390
Gene Name
AHA1, activator of heat shock protein ATPase 2
Gene Aliases
-
Chromosome Location
Chr.11: 23487882 - 23498039 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290654.1 NP_001277583.1 3 - 4 534 112
NM_001290655.1 NP_001277584.1 3 - 4 539 112
NM_172391.4 NP_765979.3 3 - 4 539 112
GenBank mRNA AK033696.1 - 3 - 4 447 112
AK035941.1 - 3 - 4 312 112
AK093945.1 - 3 - 4 446 112
AK137116.1 - 3 - 4 450 112
AK154196.1 - 3 - 4 450 112
AK155265.1 - 3 - 4 450 112
BC038397.1 - 3 - 4 425 112
BC052829.1 - 3 - 4 457 112
BC055873.1 - 3 - 4 401 112
BC064012.1 - 3 - 4 359 112

View Full Product Details