Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mfsd9
- Entrez Gene ID
-
211798
- Gene Name
- major facilitator superfamily domain containing 9
- Gene Aliases
-
4931419K03Rik
- Chromosome Location
-
Chr.1: 40772040 - 40790657 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK036610.1 |
- |
- |
2713 |
73 |
View Full Product Details