Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Kcng3
- Entrez Gene ID
-
225030
- Gene Name
- potassium voltage-gated channel, subfamily G, member 3
- Gene Aliases
-
KV6.3, Kv10.1a, Kv10.1b
- Chromosome Location
-
Chr.17: 83585411 - 83631993 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF454551.1 |
- |
1 - 2 |
900 |
71 |
View Full Product Details