Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ndufaf3
Entrez Gene ID
66706
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 3
Gene Aliases
4733401H18Rik
Chromosome Location
Chr.9: 108565865 - 108567342 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023247.1 NP_075736.1 4 - 5 865 94
GenBank mRNA AF235017.1 - 4 - 5 865 94
AK005252.1 - 4 - 5 480 94
AK014634.1 - 4 - 5 480 94
BC027513.1 - 4 - 5 898 94

View Full Product Details