The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011790.4 | NP_035920.1 | 12 - 13 | 1409 | 76 |
GenBank mRNA | AF124664.1 | - | 11 - 12 | 1236 | 76 |
AJ130975.1 | - | 11 - 12 | 1268 | 76 | |
AK011094.1 | - | 12 - 13 | 1416 | 76 | |
AK045012.1 | - | - | 1392 | 76 | |
AK149936.1 | - | 12 - 13 | 1389 | 76 | |
AK167104.1 | - | 12 - 13 | 1366 | 76 | |
AK168055.1 | - | 12 - 13 | 1412 | 76 | |
BC051998.1 | - | 12 - 13 | 1409 | 76 | |
BC052422.1 | - | 12 - 13 | 1409 | 76 |