Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h12b
Entrez Gene ID
547176
Gene Name
zinc finger CCCH-type containing 12B
Gene Aliases
A130028J20Rik, CXorf32, Gm373
Chromosome Location
Chr.X: 95711529 - 95932656 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034907.2 NP_001030079.2 8 - 9 1943 69

View Full Product Details