The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100451.2 | NP_001093921.1 | - | 656 | 75 |
GenBank mRNA | BC070432.1 | - | - | 656 | 75 |