Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Esyt3
Entrez Gene ID
272636
Gene Name
extended synaptotagmin-like protein 3
Gene Aliases
D930024E11, D9Ertd280e, Fam62c, mKIAA4186
Chromosome Location
Chr.9: 99308739 - 99360405 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177775.3 NP_808443.2 12 - 13 1457 107
GenBank mRNA AK086367.1 - 12 - 13 1458 107
AK220532.1 - 12 - 13 1445 107
BC118967.2 - - 1405 107

View Full Product Details