The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029103.3 | NP_083379.2 | 3 - 4 | 395 | 69 |
GenBank mRNA | AB009291.1 | - | 3 - 4 | 369 | 69 |
AK131997.1 | - | 3 - 4 | 396 | 69 | |
AK165646.1 | - | 3 - 4 | 371 | 69 | |
AK167580.1 | - | 3 - 4 | 395 | 69 | |
BC022761.1 | - | 3 - 4 | 401 | 69 | |
BC038901.1 | - | 3 - 4 | 383 | 69 | |
BC048616.1 | - | 2 - 3 | 458 | 69 |