Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grm2
Entrez Gene ID
108068
Gene Name
glutamate receptor, metabotropic 2
Gene Aliases
4930441L02Rik, Gprc1b, mGluR2, mGluR7
Chromosome Location
Chr.9: 106644534 - 106656109 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160353.1 NP_001153825.1 3 - 4 1508 65
GenBank mRNA BC115866.1 - 3 - 4 1481 65

View Full Product Details