The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027354.2 | NP_081630.2 | 10 - 11 | 1497 | 55 |
GenBank mRNA | AK081217.1 | - | - | 682 | 55 |
AK154163.1 | - | 10 - 11 | 1247 | 55 | |
BC034901.1 | - | 10 - 11 | 1157 | 55 | |
BE981233.1 | - | 1 - 2 | 55 |