Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Glyctk
Entrez Gene ID
235582
Gene Name
glycerate kinase
Gene Aliases
6230410P16Rik, C430006L12
Chromosome Location
Chr.9: 106152860 - 106161065 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039586.1 NP_001034675.1 4 - 5 839 90
NM_174846.4 NP_777271.3 3 - 4 975 90
GenBank mRNA AF455785.1 - 4 - 5 808 90
AK050182.1 - 4 - 5 829 90
AK052709.1 - 3 - 4 914 90
BC025834.1 - 3 - 4 944 90
BC025935.1 - - 936 90
BC030732.1 - 3 - 4 657 90
BC030736.1 - 2 - 3 224 90
BC033063.1 - 3 - 4 951 90
BC036136.1 - - 796 90

View Full Product Details