Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srbd1
Entrez Gene ID
78586
Gene Name
S1 RNA binding domain 1
Gene Aliases
AI461933, C85414, D530025C17Rik
Chromosome Location
Chr.17: 85984665 - 86145216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030133.3 NP_084409.3 5 - 6 852 151

View Full Product Details