Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Epas1
Entrez Gene ID
13819
Gene Name
endothelial PAS domain protein 1
Gene Aliases
HIF-2alpha, HIF2A, HLF, HRF, MOP2, bHLHe73
Chromosome Location
Chr.17: 86753864 - 86833410 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010137.3 NP_034267.3 15 - 16 2889 96
GenBank mRNA AF045160.1 - 15 - 16 2653 96
AK136911.1 - 15 - 16 2893 96
AK158430.1 - 15 - 16 3053 96
BC057870.1 - 15 - 16 2611 96
D89787.1 - 15 - 16 2851 96
U81983.1 - 15 - 16 2572 96

View Full Product Details