The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153597.2 | NP_705825.2 | 1 - 2 | 398 | 79 |
NR_027351.1 | 1 - 2 | 398 | 79 | ||
GenBank mRNA | AK038830.1 | - | 1 - 2 | 400 | 79 |
AK090183.1 | - | 1 - 2 | 371 | 79 | |
AK141582.1 | - | - | 355 | 79 | |
BC027575.1 | - | 1 - 2 | 196 | 79 |