Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fermt3
Entrez Gene ID
108101
Gene Name
fermitin family homolog 3 (Drosophila)
Gene Aliases
C79673, Kindlin3
Chromosome Location
Chr.19: 6998958 - 7019469 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153795.2 NP_722490.1 1 - 2 193 101
GenBank mRNA AK142912.1 - 1 - 2 194 101
AK149737.1 - 1 - 2 124 101
AK169682.1 - 1 - 2 104 101
BC025119.1 - 1 - 2 125 101

View Full Product Details