Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Naa40
Entrez Gene ID
70999
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae)
Gene Aliases
4931433E08Rik, AU023197, Nat11
Chromosome Location
Chr.19: 7225668 - 7241222 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027643.1 NP_081919.1 5 - 6 681 84
GenBank mRNA AK016504.1 - 5 - 6 642 84
AK033510.1 - 5 - 6 503 84
AK033896.1 - 5 - 6 512 84
AK035418.1 - 4 - 5 1399 84
AK162625.1 - 5 - 6 515 84
AK170548.1 - 5 - 6 507 84
BC020020.1 - 5 - 6 532 84

View Full Product Details