The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008431.2 | NP_032457.1 | 6 - 7 | 1050 | 64 |
GenBank mRNA | AF056492.1 | - | 6 - 7 | 1088 | 64 |
BC119784.1 | - | 6 - 7 | 920 | 64 |