The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026570.4 | NP_080846.1 | 1 - 2 | 249 | 68 |
GenBank mRNA | AK010522.1 | - | 1 - 2 | 250 | 68 |
AK012096.1 | - | 1 - 2 | 216 | 68 | |
AK081867.1 | - | 1 - 2 | 232 | 68 | |
BC020043.1 | - | 1 - 2 | 217 | 68 |