The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010914.2 | NP_035044.1 | 3 - 4 | 308 | 144 |
GenBank mRNA | AK010762.1 | - | 3 - 4 | 288 | 144 |
AK011792.1 | - | 3 - 4 | 227 | 144 | |
AK079209.1 | - | 3 - 4 | 263 | 144 | |
AK146168.1 | - | 3 - 4 | 310 | 144 | |
AK154749.1 | - | 3 - 4 | 270 | 144 | |
AK155749.1 | - | 3 - 4 | 270 | 144 | |
BC010719.1 | - | 3 - 4 | 249 | 144 | |
X55316.1 | - | 3 - 4 | 304 | 144 |