The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025651.4 | NP_079927.2 | 2 - 3 | 1588 | 90 |
GenBank mRNA | AK030661.1 | - | 2 - 3 | 1459 | 90 |
BC048866.1 | - | 1 - 2 | 49 | 90 | |
BC058377.1 | - | 2 - 3 | 1588 | 90 | |
BC098471.1 | - | 3 - 4 | 1473 | 90 |