The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110791.1 | NP_001104261.1 | 1 - 2 | 431 | 105 |
NM_011750.2 | NP_035880.2 | 1 - 2 | 431 | 105 | |
NR_122118.1 | 1 - 2 | 431 | 105 | ||
GenBank mRNA | AK146151.1 | - | 1 - 2 | 192 | 105 |
AK147081.1 | - | 1 - 2 | 200 | 105 | |
AK152914.1 | - | 1 - 2 | 209 | 105 | |
AK154705.1 | - | 1 - 2 | 200 | 105 | |
AK157845.1 | - | 1 - 2 | 200 | 105 | |
BC009091.1 | - | 1 - 2 | 176 | 105 | |
BC055370.1 | - | 1 - 2 | 190 | 105 | |
CJ175598.1 | - | 1 - 2 | 432 | 105 | |
X85802.1 | - | 1 - 2 | 188 | 105 |