The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028782.2 | NP_083058.2 | 10 - 11 | 1680 | 102 |
GenBank mRNA | AK004820.1 | - | 10 - 11 | 1664 | 102 |
AK131910.1 | - | 10 - 11 | 1666 | 102 | |
AK157474.1 | - | 10 - 11 | 1659 | 102 | |
AK159302.1 | - | 10 - 11 | 1665 | 102 | |
AK160071.1 | - | 10 - 11 | 1665 | 102 | |
AK160076.1 | - | 10 - 11 | 1666 | 102 | |
AK161983.1 | - | 10 - 11 | 1665 | 102 | |
AY190302.1 | - | 10 - 11 | 1650 | 102 | |
BC057365.1 | - | 8 - 9 | 3074 | 102 | |
BC058735.1 | - | 8 - 9 | 3074 | 102 | |
BC092212.1 | - | 1 - 2 | 47 | 102 |