Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rfx2
Entrez Gene ID
19725
Gene Name
regulatory factor X, 2 (influences HLA class II expression)
Gene Aliases
5430432H19Rik
Chromosome Location
Chr.17: 56775897 - 56831054 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009056.2 NP_033082.1 16 - 17 2106 69
NM_027787.1 NP_082063.1 17 - 18 2181 69
GenBank mRNA AK044439.1 - 17 - 18 2181 69
BC004654.1 - 16 - 17 2123 69
X76089.1 - 16 - 17 2061 69

View Full Product Details