Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mpp7
Entrez Gene ID
75739
Gene Name
membrane protein, palmitoylated 7 (MAGUK p55 subfamily member 7)
Gene Aliases
1110068J02Rik, 2810038M04Rik, 5430426E14Rik, AI415104, Gm955
Chromosome Location
Chr.18: 7347962 - 7626894 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081287.2 NP_001074756.2 11 - 12 1188 151
NM_001161620.1 NP_001155092.1 11 - 12 1188 151
GenBank mRNA AK036956.1 - 11 - 12 1159 151
AK078849.1 - 11 - 12 1187 151
BC117550.1 - 10 - 11 1035 151
BC118058.1 - 10 - 11 1032 148

View Full Product Details