Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Capn13
Entrez Gene ID
381122
Gene Name
calpain 13
Gene Aliases
CANP 13, Gm943
Chromosome Location
Chr.17: 73306456 - 73399306 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033444.2 NP_001028616.1 20 - 21 2073 74
GenBank mRNA AK136569.1 - 20 - 21 2074 74
BC147510.1 - 20 - 21 1990 74
BC147512.1 - 20 - 21 1990 74

View Full Product Details