Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
L3mbtl4
Entrez Gene ID
320858
Gene Name
l(3)mbt-like 4 (Drosophila)
Gene Aliases
A730037L19Rik, D930040M24Rik
Chromosome Location
Chr.17: 68273246 - 68784078 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177278.5 NP_796252.2 19 - 20 2223 61

View Full Product Details