The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003911.2 | NP_001003911.2 | 23 - 24 | 4958 | 72 |
GenBank mRNA | AK052239.1 | - | 2 - 3 | 1114 | 72 |
AL359935.1 | - | 5 - 6 | 1204 | 72 | |
BC030442.1 | - | 11 - 12 | 1399 | 72 | |
BC058991.1 | - | 20 - 21 | 3930 | 72 | |
BC076575.1 | - | 22 - 23 | 4666 | 72 | |
BC141173.1 | - | 23 - 24 | 4734 | 72 | |
BC145491.1 | - | 24 - 25 | 4557 | 72 |