Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sptbn2
Entrez Gene ID
20743
Gene Name
spectrin beta, non-erythrocytic 2
Gene Aliases
Spnb3, mKIAA0302
Chromosome Location
Chr.19: 4711223 - 4752360 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021287.1 NP_067262.1 29 - 30 6480 68
GenBank mRNA AK147671.1 - 29 - 30 6496 68
AK172931.1 - 19 - 20 4789 68
BC048851.1 - 7 - 8 1298 68
BC079628.1 - 7 - 8 1432 68
BC079860.1 - 29 - 30 6480 68

View Full Product Details