Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctsf
Entrez Gene ID
56464
Gene Name
cathepsin F
Gene Aliases
AI481912
Chromosome Location
Chr.19: 4855129 - 4860912 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019861.1 NP_063914.1 1 - 2 361 75
GenBank mRNA AF136280.1 - 1 - 2 262 75
AF197480.1 - 1 - 2 306 75
AK017474.1 - 1 - 2 247 74
AK075862.1 - 1 - 2 254 75
AK165416.1 - 1 - 2 329 75
BC058758.1 - 1 - 2 252 75

View Full Product Details