Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adrbk1
Entrez Gene ID
110355
Gene Name
adrenergic receptor kinase, beta 1
Gene Aliases
Adrbk-1, Bark-1, GRK2, beta ARK, betaARK1
Chromosome Location
Chr.19: 4285999 - 4306222 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290818.1 NP_001277747.1 11 - 12 1167 74
NM_130863.2 NP_570933.1 12 - 13 1239 74
GenBank mRNA AF333028.1 - 11 - 12 959 74
AK155691.1 - 11 - 12 1168 74
BC003196.1 - 11 - 12 955 74
BC033272.1 - 10 - 11 1283 74
BC053922.1 - 12 - 13 972 74

View Full Product Details