Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bbs1
Entrez Gene ID
52028
Gene Name
Bardet-Biedl syndrome 1 (human)
Gene Aliases
AI451249, D19Ertd609e
Chromosome Location
Chr.19: 4886878 - 4906627 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033128.3 NP_001028300.1 8 - 9 745 63
GenBank mRNA AK037753.1 - 8 - 9 731 63
AK045850.1 - 8 - 9 743 63
AK161343.1 - 8 - 9 739 63
BC076577.1 - 8 - 9 734 63

View Full Product Details